Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609671 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STEAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STEAP3 - STEAP3 metalloreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008410.1 | Intron | NP_001008410.1 | ||||
NM_018234.2 | Intron | NP_060704.2 | ||||
NM_138637.2 | Intron | NP_619543.2 | ||||
NM_182915.2 | Intron | NP_878919.2 | ||||
XM_006712614.3 | Intron | XP_006712677.1 | ||||
XM_006712615.1 | Intron | XP_006712678.1 | ||||
XM_011511403.1 | Intron | XP_011509705.1 |