Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609671 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STEAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STEAP3 - STEAP3 metalloreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008410.1 | Intron | NP_001008410.1 | ||||
NM_018234.2 | Intron | NP_060704.2 | ||||
NM_138637.2 | Intron | NP_619543.2 | ||||
NM_182915.2 | Intron | NP_878919.2 | ||||
XM_006712614.3 | Intron | XP_006712677.1 | ||||
XM_006712615.1 | Intron | XP_006712678.1 | ||||
XM_011511403.1 | Intron | XP_011509705.1 |