Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182098 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCTR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SCTR - secretin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002980.2 | 775 | Silent Mutation | AAC,AAT | N,N 392 | NP_002971.2 | |
XM_005263730.4 | 775 | Silent Mutation | AAC,AAT | N,N 214 | XP_005263787.1 | |
XM_011511621.2 | 775 | Silent Mutation | AAC,AAT | N,N 397 | XP_011509923.1 | |
XM_017004670.1 | 775 | Silent Mutation | AAC,AAT | N,N 387 | XP_016860159.1 | |
XM_017004671.1 | 775 | Silent Mutation | AAC,AAT | N,N 361 | XP_016860160.1 | |
XM_017004672.1 | 775 | Silent Mutation | AAC,AAT | N,N 329 | XP_016860161.1 | |
XM_017004673.1 | 775 | Silent Mutation | AAC,AAT | N,N 219 | XP_016860162.1 |
TMEM37 - transmembrane protein 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |