Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606731 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OSBPL2 - oxysterol binding protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278649.1 | Intron | NP_001265578.1 | ||||
NM_014835.3 | Intron | NP_055650.1 | ||||
NM_144498.2 | Intron | NP_653081.1 | ||||
XM_017028163.1 | Intron | XP_016883652.1 | ||||
XM_017028164.1 | Intron | XP_016883653.1 | ||||
XM_017028165.1 | Intron | XP_016883654.1 | ||||
XM_017028166.1 | Intron | XP_016883655.1 | ||||
XM_017028167.1 | Intron | XP_016883656.1 | ||||
XM_017028168.1 | Intron | XP_016883657.1 | ||||
XM_017028169.1 | Intron | XP_016883658.1 | ||||
XM_017028170.1 | Intron | XP_016883659.1 |