Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGAGGCAGGGCACTCATGGGTC[A/G]CTTCTGGAGATGTCCACATTTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606731 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369209 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105369209 - putative uncharacterized protein FLJ44790 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011529124.1 | 1632 | UTR 3 | XP_011527426.1 |
OSBPL2 - oxysterol binding protein like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |