Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GUCD1 - guanylyl cyclase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284251.1 | 1313 | UTR 3 | NP_001271180.1 | |||
NM_001284252.1 | 1313 | UTR 3 | NP_001271181.1 | |||
NM_001284253.1 | 1313 | UTR 3 | NP_001271182.1 | |||
NM_001284254.1 | 1313 | UTR 3 | NP_001271183.1 | |||
NM_001284255.1 | 1313 | UTR 3 | NP_001271184.1 | |||
NM_001284256.1 | 1313 | UTR 3 | NP_001271185.1 | |||
NM_001284257.1 | 1313 | UTR 3 | NP_001271186.1 | |||
NM_031444.3 | 1313 | UTR 3 | NP_113632.2 | |||
XM_005261761.1 | 1313 | UTR 3 | XP_005261818.1 |
Set Membership: |
HapMap Validated |