Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 608515 MIM: 610964 | ||||||||||||||||||||||||||
Literature Links: |
NCF2 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NCF2 - neutrophil cytosolic factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000433.3 | Intron | NP_000424.2 | ||||
NM_001127651.2 | Intron | NP_001121123.1 | ||||
NM_001190789.1 | Intron | NP_001177718.1 | ||||
NM_001190794.1 | Intron | NP_001177723.1 | ||||
XM_005245207.1 | Intron | XP_005245264.1 | ||||
XM_011509580.1 | Intron | XP_011507882.1 | ||||
XM_011509581.1 | Intron | XP_011507883.1 |
SMG7 - SMG7, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |