Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615507 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NISCH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NISCH - nischarin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276293.1 | Intron | NP_001263222.1 | ||||
NM_001276294.1 | Intron | NP_001263223.1 | ||||
NM_007184.3 | Intron | NP_009115.2 | ||||
XM_005264839.2 | Intron | XP_005264896.1 | ||||
XM_006712955.2 | Intron | XP_006713018.1 | ||||
XM_017005643.1 | Intron | XP_016861132.1 |
Set Membership: |
HapMap |