Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601865 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLOD2 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000935.2 | 2377 | UTR 3 | NP_000926.2 | |||
NM_182943.2 | 2377 | UTR 3 | NP_891988.1 | |||
XM_005247535.3 | 2377 | UTR 3 | XP_005247592.1 | |||
XM_017006625.1 | 2377 | UTR 3 | XP_016862114.1 |
Set Membership: |
HapMap |