Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610421 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHDRBS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KHDRBS3 - KH RNA binding domain containing, signal transduction associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006558.2 | Intron | NP_006549.1 | ||||
XM_005250757.2 | Intron | XP_005250814.1 | ||||
XM_005250758.3 | Intron | XP_005250815.1 | ||||
XM_011516796.2 | Intron | XP_011515098.1 | ||||
XM_011516797.2 | Intron | XP_011515099.1 | ||||
XM_011516798.1 | Intron | XP_011515100.1 | ||||
XM_011516799.1 | Intron | XP_011515101.1 |
Set Membership: |
HapMap |