Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601970 | ||||||||||||||||||||
Literature Links: |
LINC00689 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00689 - long intergenic non-protein coding RNA 689 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIPR2 - vasoactive intestinal peptide receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304522.1 | 1502 | Missense Mutation | CAC,CGC | H,R 332 | NP_001291451.1 | |
NM_001308259.1 | 1502 | Missense Mutation | CAC,CGC | H,R 396 | NP_001295188.1 | |
NM_003382.4 | 1502 | Missense Mutation | CAC,CGC | H,R 412 | NP_003373.2 | |
XM_005249561.3 | 1502 | Missense Mutation | CAC,CGC | H,R 437 | XP_005249618.1 | |
XM_006716107.2 | 1502 | Intron | XP_006716170.1 | |||
XM_006716108.3 | 1502 | Intron | XP_006716171.1 | |||
XM_011516550.2 | 1502 | Intron | XP_011514852.1 | |||
XM_017012580.1 | 1502 | Missense Mutation | CAC,CGC | H,R 274 | XP_016868069.1 |