Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612377 | ||||||||||||||||||||
Literature Links: |
COMMD6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COMMD6 - COMM domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287392.1 | 307 | Silent Mutation | GTG,GTT | V,V 22 | NP_001274321.1 | |
NM_001287393.1 | 307 | Silent Mutation | GTG,GTT | V,V 22 | NP_001274322.1 | |
NM_001287394.1 | 307 | Silent Mutation | GTG,GTT | V,V 22 | NP_001274323.1 | |
NM_203495.3 | 307 | Silent Mutation | GTG,GTT | V,V 49 | NP_987091.1 | |
NM_203497.3 | 307 | Silent Mutation | GTG,GTT | V,V 49 | NP_987093.1 | |
XM_011534948.2 | 307 | Silent Mutation | GTG,GTT | V,V 89 | XP_011533250.1 |