Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611956 MIM: 607792 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C3orf52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C3orf52 - chromosome 3 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171747.1 | 1836 | Intron | NP_001165218.1 | |||
NM_024616.2 | 1836 | Intron | NP_078892.2 | |||
XM_011513133.2 | 1836 | Intron | XP_011511435.1 | |||
XM_017007181.1 | 1836 | Intron | XP_016862670.1 |
GCSAM - germinal center associated signaling and motility | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190259.1 | 1836 | UTR 3 | NP_001177188.1 | |||
NM_001190260.1 | 1836 | UTR 3 | NP_001177189.1 | |||
NM_152785.4 | 1836 | UTR 3 | NP_689998.1 | |||
XM_005247272.2 | 1836 | UTR 3 | XP_005247329.1 | |||
XM_011512627.2 | 1836 | UTR 3 | XP_011510929.1 | |||
XM_011512628.2 | 1836 | UTR 3 | XP_011510930.1 | |||
XM_011512629.1 | 1836 | UTR 3 | XP_011510931.1 | |||
XM_011512630.1 | 1836 | UTR 3 | XP_011510932.1 |
MIR567 - microRNA 567 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |