Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605139 MIM: 607743 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCT2 - chaperonin containing TCP1 subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198842.1 | Intron | NP_001185771.1 | ||||
NM_006431.2 | Intron | NP_006422.1 |
FRS2 - fibroblast growth factor receptor substrate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3913-1 - microRNA 3913-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3913-2 - microRNA 3913-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |