Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEKT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEKT3 - tektin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031898.2 | 2965 | Silent Mutation | NP_114104.1 | |||
XM_011523988.2 | 2965 | Silent Mutation | XP_011522290.1 | |||
XM_011523989.1 | 2965 | Silent Mutation | XP_011522291.1 | |||
XM_011523990.2 | 2965 | Silent Mutation | XP_011522292.1 | |||
XM_017024954.1 | 2965 | Silent Mutation | XP_016880443.1 | |||
XM_017024955.1 | 2965 | Silent Mutation | XP_016880444.1 | |||
XM_017024956.1 | 2965 | Intron | XP_016880445.1 |