Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601097 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMP22 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PMP22 - peripheral myelin protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000304.3 | 910 | UTR 3 | NP_000295.1 | |||
NM_001281455.1 | 910 | UTR 3 | NP_001268384.1 | |||
NM_001281456.1 | 910 | UTR 3 | NP_001268385.1 | |||
NM_153321.2 | 910 | UTR 3 | NP_696996.1 | |||
NM_153322.2 | 910 | UTR 3 | NP_696997.1 | |||
XM_017024775.1 | 910 | Intron | XP_016880264.1 | |||
XM_017024776.1 | 910 | Intron | XP_016880265.1 |
Set Membership: |
HapMap |