Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609896 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EIF4E3 - eukaryotic translation initiation factor 4E family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134649.2 | Intron | NP_001128121.1 | ||||
NM_001134650.1 | Intron | NP_001128122.1 | ||||
NM_001134651.1 | Intron | NP_001128123.1 | ||||
NM_001282886.1 | Intron | NP_001269815.1 | ||||
NM_173359.4 | Intron | NP_775495.1 | ||||
XM_011533651.2 | Intron | XP_011531953.1 | ||||
XM_017006278.1 | Intron | XP_016861767.1 | ||||
XM_017006279.1 | Intron | XP_016861768.1 | ||||
XM_017006280.1 | Intron | XP_016861769.1 | ||||
XM_017006281.1 | Intron | XP_016861770.1 | ||||
XM_017006282.1 | Intron | XP_016861771.1 |
Set Membership: |
HapMap Validated |