Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTAGCACCTAACATAGTTTAT[G/A]GGGTATTTTCCAGTCATTTCATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606127 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928418 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928418 - uncharacterized LOC101928418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYOCD - myocardin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146312.2 | Intron | NP_001139784.1 | ||||
NM_153604.3 | Intron | NP_705832.1 | ||||
XM_005256863.1 | Intron | XP_005256920.1 | ||||
XM_005256864.1 | Intron | XP_005256921.1 | ||||
XM_017025342.1 | Intron | XP_016880831.1 |
Set Membership: |
HapMap |