Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180202 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC77 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC77 - coiled-coil domain containing 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130146.1 | Intron | NP_001123618.1 | ||||
NM_001130147.1 | Intron | NP_001123619.1 | ||||
NM_001130148.1 | Intron | NP_001123620.1 | ||||
NM_032358.3 | Intron | NP_115734.1 |
KDM5A - lysine demethylase 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |