Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180202 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC77 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC77 - coiled-coil domain containing 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130146.1 | Intron | NP_001123618.1 | ||||
NM_001130147.1 | Intron | NP_001123619.1 | ||||
NM_001130148.1 | Intron | NP_001123620.1 | ||||
NM_032358.3 | Intron | NP_115734.1 |
KDM5A - lysine demethylase 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |