Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158380 MIM: 158381 MIM: 613113 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVI2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EVI2A - ecotropic viral integration site 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003927.2 | 1445 | UTR 3 | NP_001003927.1 | |||
NM_014210.3 | 1445 | UTR 3 | NP_055025.2 |
EVI2B - ecotropic viral integration site 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NF1 - neurofibromin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000267.3 | 1445 | Intron | NP_000258.1 | |||
NM_001042492.2 | 1445 | Intron | NP_001035957.1 | |||
NM_001128147.2 | 1445 | Intron | NP_001121619.1 | |||
XM_005257983.2 | 1445 | Intron | XP_005258040.1 | |||
XM_005257984.2 | 1445 | Intron | XP_005258041.1 | |||
XM_006721922.2 | 1445 | Intron | XP_006721985.1 | |||
XM_006721924.2 | 1445 | Intron | XP_006721987.1 | |||
XM_006721925.2 | 1445 | Intron | XP_006721988.1 | |||
XM_006721926.3 | 1445 | Intron | XP_006721989.1 | |||
XM_011524852.2 | 1445 | Intron | XP_011523154.1 | |||
XM_011524857.2 | 1445 | Intron | XP_011523159.1 | |||
XM_017024689.1 | 1445 | Intron | XP_016880178.1 | |||
XM_017024690.1 | 1445 | Intron | XP_016880179.1 |
Set Membership: |
HapMap Validated |