Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602843 MIM: 610642 | ||||||||||||||||||||
Literature Links: |
ARHGDIB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGDIB - Rho GDP dissociation inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001175.6 | 1168 | UTR 3 | NP_001166.3 | |||
NM_001321420.1 | 1168 | UTR 3 | NP_001308349.1 | |||
NM_001321421.1 | 1168 | UTR 3 | NP_001308350.1 | |||
NM_001321422.1 | 1168 | UTR 3 | NP_001308351.1 | |||
NM_001321423.1 | 1168 | UTR 3 | NP_001308352.1 |
ERP27 - endoplasmic reticulum protein 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |