Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612141 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPB41L4A - erythrocyte membrane protein band 4.1 like 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022140.3 | Intron | NP_071423.3 | ||||
XM_011543530.1 | Intron | XP_011541832.1 | ||||
XM_011543531.2 | Intron | XP_011541833.1 | ||||
XM_011543532.1 | Intron | XP_011541834.1 | ||||
XM_011543533.1 | Intron | XP_011541835.1 | ||||
XM_017009689.1 | Intron | XP_016865178.1 |