Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612141 | |||||||||||||||||||||||
Literature Links: |
EPB41L4A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
EPB41L4A - erythrocyte membrane protein band 4.1 like 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022140.3 | 3741 | Intron | NP_071423.3 | |||
XM_011543530.1 | 3741 | UTR 3 | XP_011541832.1 | |||
XM_011543531.2 | 3741 | Intron | XP_011541833.1 | |||
XM_011543532.1 | 3741 | UTR 3 | XP_011541834.1 | |||
XM_011543533.1 | 3741 | UTR 3 | XP_011541835.1 | |||
XM_017009689.1 | 3741 | Intron | XP_016865178.1 |