Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 109530 | ||||||||||||||||||||
Literature Links: |
CD48 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD48 - CD48 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256030.1 | 959 | Intron | NP_001242959.1 | |||
NM_001778.3 | 959 | UTR 3 | NP_001769.2 | |||
XM_005245625.1 | 959 | UTR 3 | XP_005245682.1 | |||
XM_011510171.1 | 959 | UTR 3 | XP_011508473.1 | |||
XM_017002867.1 | 959 | UTR 3 | XP_016858356.1 |