Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
78 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109530 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD48 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD48 - CD48 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256030.1 | 1181 | Intron | NP_001242959.1 | |||
NM_001778.3 | 1181 | UTR 3 | NP_001769.2 | |||
XM_005245625.1 | 1181 | UTR 3 | XP_005245682.1 | |||
XM_011510171.1 | 1181 | UTR 3 | XP_011508473.1 | |||
XM_017002867.1 | 1181 | UTR 3 | XP_016858356.1 |
Set Membership: |
HapMap JSNP |