Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602948 MIM: 612012 | ||||||||||||||||||||
Literature Links: |
RAD51B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD51B - RAD51 paralog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321809.1 | Intron | NP_001308738.1 | ||||
NM_001321810.1 | Intron | NP_001308739.1 | ||||
NM_001321812.1 | Intron | NP_001308741.1 | ||||
NM_001321814.1 | Intron | NP_001308743.1 | ||||
NM_001321815.1 | Intron | NP_001308744.1 | ||||
NM_001321817.1 | Intron | NP_001308746.1 | ||||
NM_001321818.1 | Intron | NP_001308747.1 | ||||
NM_001321819.1 | Intron | NP_001308748.1 | ||||
NM_001321821.1 | Intron | NP_001308750.1 | ||||
NM_002877.5 | Intron | NP_002868.1 | ||||
NM_133509.3 | Intron | NP_598193.2 | ||||
NM_133510.3 | Intron | NP_598194.1 | ||||
XM_011537050.2 | Intron | XP_011535352.1 | ||||
XM_011537051.2 | Intron | XP_011535353.1 | ||||
XM_017021545.1 | Intron | XP_016877034.1 | ||||
XM_017021546.1 | Intron | XP_016877035.1 | ||||
XM_017021547.1 | Intron | XP_016877036.1 | ||||
XM_017021548.1 | Intron | XP_016877037.1 |
ZFYVE26 - zinc finger FYVE-type containing 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |