Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601413 | ||||||||||||||||||||
Literature Links: |
DIO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIO2 - deiodinase, iodothyronine, type II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000793.5 | 5274 | UTR 3 | NP_000784.2 | |||
NM_001007023.4 | 5274 | UTR 3 | NP_001007024.1 | |||
NM_001242502.2 | 5274 | UTR 3 | NP_001229431.1 | |||
NM_001242503.2 | 5274 | UTR 3 | NP_001229432.1 | |||
NM_001324462.1 | 5274 | UTR 3 | NP_001311391.1 | |||
NM_013989.4 | 5274 | UTR 3 | NP_054644.1 |