Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137350 MIM: 133090 | ||||||||||||||||||||
Literature Links: |
GSN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSN - gelsolin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STOM - stomatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270526.1 | 2766 | UTR 3 | NP_001257455.1 | |||
NM_001270527.1 | 2766 | UTR 3 | NP_001257456.1 | |||
NM_004099.5 | 2766 | UTR 3 | NP_004090.4 | |||
NM_198194.2 | 2766 | UTR 3 | NP_937837.1 |