Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605620 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL20RA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL20RA - interleukin 20 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278722.1 | 2435 | UTR 3 | NP_001265651.1 | |||
NM_001278723.1 | 2435 | UTR 3 | NP_001265652.1 | |||
NM_001278724.1 | 2435 | UTR 3 | NP_001265653.1 | |||
NM_014432.3 | 2435 | UTR 3 | NP_055247.3 | |||
XM_006715506.2 | 2435 | UTR 3 | XP_006715569.1 | |||
XM_011535904.2 | 2435 | UTR 3 | XP_011534206.1 | |||
XM_017010954.1 | 2435 | UTR 3 | XP_016866443.1 | |||
XM_017010955.1 | 2435 | UTR 3 | XP_016866444.1 |
NHEG1 - neuroblastoma highly expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |