Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610715 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEMGN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HEMGN - hemogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018437.4 | Intron | NP_060907.2 | ||||
NM_197978.2 | Intron | NP_932095.1 | ||||
XM_005252086.1 | Intron | XP_005252143.1 | ||||
XM_011518845.2 | Intron | XP_011517147.1 | ||||
XM_011518846.2 | Intron | XP_011517148.1 |
TRMO - tRNA methyltransferase O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |