Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM161B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM161B - family with sequence similarity 161 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152445.2 | 2283 | Intron | NP_689658.2 | |||
XM_011536475.1 | 2283 | UTR 3 | XP_011534777.1 |
ZNF410 - zinc finger protein 410 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242924.1 | 2283 | UTR 3 | NP_001229853.1 | |||
NM_001242926.1 | 2283 | UTR 3 | NP_001229855.1 | |||
NM_001242927.1 | 2283 | UTR 3 | NP_001229856.1 | |||
NM_001242928.1 | 2283 | UTR 3 | NP_001229857.1 | |||
NM_021188.2 | 2283 | UTR 3 | NP_067011.1 |