Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610192 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIS3 - GLIS family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042413.1 | 5589 | UTR 3 | NP_001035878.1 | |||
NM_152629.3 | 5589 | UTR 3 | NP_689842.3 | |||
XM_005251386.4 | 5589 | UTR 3 | XP_005251443.1 | |||
XM_005251387.4 | 5589 | UTR 3 | XP_005251444.1 | |||
XM_005251388.4 | 5589 | UTR 3 | XP_005251445.1 | |||
XM_005251389.4 | 5589 | Intron | XP_005251446.1 | |||
XM_006716731.3 | 5589 | Intron | XP_006716794.1 | |||
XM_011517763.2 | 5589 | UTR 3 | XP_011516065.1 | |||
XM_011517764.2 | 5589 | UTR 3 | XP_011516066.1 | |||
XM_011517765.2 | 5589 | Intron | XP_011516067.1 | |||
XM_011517766.2 | 5589 | UTR 3 | XP_011516068.1 | |||
XM_011517767.2 | 5589 | UTR 3 | XP_011516069.1 | |||
XM_011517769.2 | 5589 | Intron | XP_011516071.1 | |||
XM_017014361.1 | 5589 | UTR 3 | XP_016869850.1 |