Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616114 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHD6 - chromodomain helicase DNA binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032221.4 | 9721 | UTR 3 | NP_115597.3 | |||
XM_005260573.2 | 9721 | UTR 3 | XP_005260630.1 | |||
XM_005260576.4 | 9721 | UTR 3 | XP_005260633.1 | |||
XM_011529080.2 | 9721 | UTR 3 | XP_011527382.1 | |||
XM_011529082.2 | 9721 | UTR 3 | XP_011527384.1 | |||
XM_017028099.1 | 9721 | UTR 3 | XP_016883588.1 | |||
XM_017028100.1 | 9721 | UTR 3 | XP_016883589.1 | |||
XM_017028101.1 | 9721 | UTR 3 | XP_016883590.1 | |||
XM_017028102.1 | 9721 | Intron | XP_016883591.1 | |||
XM_017028103.1 | 9721 | UTR 3 | XP_016883592.1 | |||
XM_017028104.1 | 9721 | Intron | XP_016883593.1 |