Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610317 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COBL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COBL - cordon-bleu WH2 repeat protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287436.1 | 4951 | UTR 3 | NP_001274365.1 | |||
NM_001287438.1 | 4951 | Intron | NP_001274367.1 | |||
NM_015198.3 | 4951 | UTR 3 | NP_056013.2 | |||
XM_005271750.1 | 4951 | UTR 3 | XP_005271807.1 | |||
XM_005271751.1 | 4951 | UTR 3 | XP_005271808.1 | |||
XM_005271756.1 | 4951 | UTR 3 | XP_005271813.1 | |||
XM_011515234.1 | 4951 | UTR 3 | XP_011513536.1 | |||
XM_011515235.1 | 4951 | UTR 3 | XP_011513537.1 | |||
XM_011515236.1 | 4951 | UTR 3 | XP_011513538.1 | |||
XM_011515237.1 | 4951 | UTR 3 | XP_011513539.1 | |||
XM_011515238.1 | 4951 | UTR 3 | XP_011513540.1 | |||
XM_011515239.1 | 4951 | UTR 3 | XP_011513541.1 | |||
XM_011515240.1 | 4951 | UTR 3 | XP_011513542.1 | |||
XM_011515241.1 | 4951 | UTR 3 | XP_011513543.1 | |||
XM_017011898.1 | 4951 | UTR 3 | XP_016867387.1 | |||
XM_017011899.1 | 4951 | UTR 3 | XP_016867388.1 | |||
XM_017011900.1 | 4951 | UTR 3 | XP_016867389.1 | |||
XM_017011901.1 | 4951 | Intron | XP_016867390.1 |