Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611108 MIM: 608498 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VWC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
VWC2 - von Willebrand factor C domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198570.4 | Intron | NP_940972.2 |
ZPBP - zona pellucida binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159878.1 | Intron | NP_001153350.1 | ||||
NM_007009.2 | Intron | NP_008940.2 | ||||
XM_011515095.2 | Intron | XP_011513397.1 | ||||
XM_011515096.2 | Intron | XP_011513398.1 | ||||
XM_011515097.1 | Intron | XP_011513399.1 | ||||
XM_011515100.2 | Intron | XP_011513402.1 | ||||
XM_011515101.2 | Intron | XP_011513403.1 | ||||
XM_011515102.2 | Intron | XP_011513404.1 | ||||
XM_011515103.1 | Intron | XP_011513405.1 | ||||
XM_017011707.1 | Intron | XP_016867196.1 |