Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609104 | ||||||||||||||||||||
Literature Links: |
FBXO34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO34 - F-box protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017943.3 | Intron | NP_060413.2 | ||||
NM_152231.1 | Intron | NP_689417.1 | ||||
XM_005267791.4 | Intron | XP_005267848.1 | ||||
XM_006720185.3 | Intron | XP_006720248.1 | ||||
XM_017021391.1 | Intron | XP_016876880.1 | ||||
XM_017021392.1 | Intron | XP_016876881.1 | ||||
XM_017021393.1 | Intron | XP_016876882.1 |