Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614798 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PELI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PELI2 - pellino E3 ubiquitin protein ligase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021255.2 | Intron | NP_067078.1 | ||||
XM_005267890.4 | Intron | XP_005267947.1 | ||||
XM_006720211.3 | Intron | XP_006720274.1 | ||||
XM_011536990.2 | Intron | XP_011535292.1 | ||||
XM_011536992.2 | Intron | XP_011535294.1 | ||||
XM_017021478.1 | Intron | XP_016876967.1 | ||||
XM_017021479.1 | Intron | XP_016876968.1 |
Set Membership: |
HapMap |