Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKI - diacylglycerol kinase iota | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321708.1 | 4522 | UTR 3 | NP_001308637.1 | |||
NM_001321709.1 | 4522 | UTR 3 | NP_001308638.1 | |||
NM_001321710.1 | 4522 | UTR 3 | NP_001308639.1 | |||
NM_004717.3 | 4522 | UTR 3 | NP_004708.1 | |||
XM_011516682.2 | 4522 | Intron | XP_011514984.2 | |||
XM_011516684.2 | 4522 | UTR 3 | XP_011514986.2 | |||
XM_017012780.1 | 4522 | UTR 3 | XP_016868269.1 | |||
XM_017012781.1 | 4522 | UTR 3 | XP_016868270.1 | |||
XM_017012782.1 | 4522 | UTR 3 | XP_016868271.1 | |||
XM_017012783.1 | 4522 | UTR 3 | XP_016868272.1 | |||
XM_017012784.1 | 4522 | UTR 3 | XP_016868273.1 | |||
XM_017012785.1 | 4522 | UTR 3 | XP_016868274.1 | |||
XM_017012786.1 | 4522 | UTR 3 | XP_016868275.1 | |||
XM_017012787.1 | 4522 | UTR 3 | XP_016868276.1 | |||
XM_017012788.1 | 4522 | UTR 3 | XP_016868277.1 | |||
XM_017012789.1 | 4522 | UTR 3 | XP_016868278.1 |