Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612990 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASXL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASXL1 - additional sex combs like 1, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOL4L - nucleolar protein 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256798.1 | 1835 | UTR 3 | NP_001243727.1 | |||
NM_080616.4 | 1835 | UTR 3 | NP_542183.2 | |||
XM_005260282.4 | 1835 | Intron | XP_005260339.1 | |||
XM_005260283.4 | 1835 | Intron | XP_005260340.1 | |||
XM_005260284.4 | 1835 | Intron | XP_005260341.1 | |||
XM_005260285.4 | 1835 | Intron | XP_005260342.2 | |||
XM_005260286.4 | 1835 | Intron | XP_005260343.1 | |||
XM_005260288.2 | 1835 | Intron | XP_005260345.1 | |||
XM_005260289.4 | 1835 | Intron | XP_005260346.1 | |||
XM_006723702.3 | 1835 | Intron | XP_006723765.1 | |||
XM_006723703.3 | 1835 | Intron | XP_006723766.1 | |||
XM_011528562.2 | 1835 | Intron | XP_011526864.1 | |||
XM_011528563.1 | 1835 | Intron | XP_011526865.1 | |||
XM_017027669.1 | 1835 | Intron | XP_016883158.1 | |||
XM_017027670.1 | 1835 | Intron | XP_016883159.1 |