Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608037 MIM: 615245 MIM: 601737 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHPF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHPF2 - chondroitin polymerizing factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR671 - microRNA 671 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCD3 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003801.1 | 918 | Silent Mutation | CAC,CAT | H,H 276 | NP_001003801.1 | |
NM_001003802.1 | 918 | Silent Mutation | CAC,CAT | H,H 263 | NP_001003802.1 | |
NM_003078.3 | 918 | Silent Mutation | CAC,CAT | H,H 263 | NP_003069.2 | |
XM_011516521.2 | 918 | Silent Mutation | CAC,CAT | H,H 174 | XP_011514823.1 | |
XM_017012556.1 | 918 | Silent Mutation | CAC,CAT | H,H 111 | XP_016868045.1 |
Set Membership: |
HapMap |