Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612510 | ||||||||||||||||||||
Literature Links: |
ABCF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCF2 - ATP binding cassette subfamily F member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005692.4 | 1988 | Silent Mutation | NP_005683.2 | |||
NM_007189.2 | 1988 | Silent Mutation | NP_009120.1 | |||
XM_017011639.1 | 1988 | Silent Mutation | XP_016867128.1 | |||
XM_017011640.1 | 1988 | Missense Mutation | XP_016867129.1 |
IQCA1L - IQ motif containing with AAA domain 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |