Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163729 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOS3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOS3 - nitric oxide synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000603.4 | Intron | NP_000594.2 | ||||
NM_001160109.1 | Intron | NP_001153581.1 | ||||
NM_001160110.1 | Intron | NP_001153582.1 | ||||
NM_001160111.1 | Intron | NP_001153583.1 | ||||
XM_006716002.3 | Intron | XP_006716065.1 | ||||
XM_017012232.1 | Intron | XP_016867721.1 | ||||
XM_017012233.1 | Intron | XP_016867722.1 | ||||
XM_017012234.1 | Intron | XP_016867723.1 |
Set Membership: |
HapMap |