Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM221A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM221A - family with sequence similarity 221 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK31 - serine/threonine kinase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260504.1 | 269 | UTR 5 | NP_001247433.1 | |||
NM_001260505.1 | 269 | Intron | NP_001247434.1 | |||
NM_031414.4 | 269 | Intron | NP_113602.2 | |||
NM_032944.3 | 269 | UTR 5 | NP_116562.2 | |||
XM_006715758.3 | 269 | Intron | XP_006715821.1 | |||
XM_011515449.2 | 269 | Intron | XP_011513751.2 | |||
XM_011515450.1 | 269 | UTR 5 | XP_011513752.1 | |||
XM_011515452.2 | 269 | Intron | XP_011513754.2 |