Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCCCTGTCATATCACTACATC[C/T]GATTCTCTCAAATGTTATAAAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612045 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1QTNF3 - C1q and tumor necrosis factor related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030945.3 | 1512 | UTR 3 | NP_112207.1 | |||
NM_181435.5 | 1512 | UTR 3 | NP_852100.3 |
C1QTNF3-AMACR - C1QTNF3-AMACR readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |