Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VWA5B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VWA5B1 - von Willebrand factor A domain containing 5B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039500.2 | Intron | NP_001034589.2 | ||||
XM_006710360.3 | Intron | XP_006710423.2 | ||||
XM_011540683.2 | Intron | XP_011538985.1 | ||||
XM_011540684.2 | Intron | XP_011538986.1 | ||||
XM_011540685.2 | Intron | XP_011538987.1 | ||||
XM_011540688.2 | Intron | XP_011538990.1 | ||||
XM_011540690.2 | Intron | XP_011538992.1 | ||||
XM_011540691.2 | Intron | XP_011538993.1 | ||||
XM_011540692.2 | Intron | XP_011538994.1 | ||||
XM_011540693.1 | Intron | XP_011538995.1 | ||||
XM_011540695.1 | Intron | XP_011538997.1 | ||||
XM_011540697.2 | Intron | XP_011538999.1 | ||||
XM_011540698.1 | Intron | XP_011539000.1 | ||||
XM_017000305.1 | Intron | XP_016855794.1 | ||||
XM_017000306.1 | Intron | XP_016855795.1 |
Set Membership: |
HapMap |