Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
73 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601192 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PLA2G5 - phospholipase A2 group V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000929.2 | Intron | NP_000920.1 | ||||
XM_005245891.4 | Intron | XP_005245948.1 | ||||
XM_005245892.4 | Intron | XP_005245949.1 | ||||
XM_005245893.4 | Intron | XP_005245950.1 | ||||
XM_011541586.2 | Intron | XP_011539888.1 | ||||
XM_011541587.2 | Intron | XP_011539889.1 | ||||
XM_011541588.2 | Intron | XP_011539890.1 | ||||
XM_011541589.2 | Intron | XP_011539891.1 | ||||
XM_011541590.2 | Intron | XP_011539892.1 | ||||
XM_011541591.2 | Intron | XP_011539893.1 | ||||
XM_011541592.2 | Intron | XP_011539894.1 |
Set Membership: |
HapMap Validated |