Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607715 | ||||||||||||||||||||
Literature Links: |
TSC22D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSC22D1 - TSC22 domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243797.1 | 633 | UTR 3 | NP_001230726.1 | |||
NM_001243798.1 | 633 | UTR 3 | NP_001230727.1 | |||
NM_001243799.1 | 633 | Intron | NP_001230728.1 | |||
NM_006022.3 | 633 | UTR 3 | NP_006013.1 | |||
NM_183422.3 | 633 | UTR 3 | NP_904358.2 | |||
XM_005266583.4 | 633 | Intron | XP_005266640.1 | |||
XM_017020810.1 | 633 | Intron | XP_016876299.1 | |||
XM_017020811.1 | 633 | Intron | XP_016876300.1 |