Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605314 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HDAC4 - histone deacetylase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006037.3 | 8314 | UTR 3 | NP_006028.2 | |||
XM_006712877.3 | 8314 | UTR 3 | XP_006712940.1 | |||
XM_006712878.3 | 8314 | UTR 3 | XP_006712941.1 | |||
XM_006712879.3 | 8314 | UTR 3 | XP_006712942.1 | |||
XM_006712880.3 | 8314 | UTR 3 | XP_006712943.1 | |||
XM_011512217.2 | 8314 | UTR 3 | XP_011510519.1 | |||
XM_011512218.2 | 8314 | UTR 3 | XP_011510520.1 | |||
XM_011512219.2 | 8314 | UTR 3 | XP_011510521.1 | |||
XM_011512220.2 | 8314 | UTR 3 | XP_011510522.1 | |||
XM_011512221.1 | 8314 | UTR 3 | XP_011510523.1 | |||
XM_011512222.2 | 8314 | UTR 3 | XP_011510524.1 | |||
XM_011512223.2 | 8314 | UTR 3 | XP_011510525.1 | |||
XM_011512224.2 | 8314 | UTR 3 | XP_011510526.1 | |||
XM_011512225.2 | 8314 | UTR 3 | XP_011510527.1 | |||
XM_011512226.2 | 8314 | UTR 3 | XP_011510528.1 | |||
XM_011512227.2 | 8314 | UTR 3 | XP_011510529.1 | |||
XM_011512230.1 | 8314 | UTR 3 | XP_011510532.1 | |||
XM_017005394.1 | 8314 | UTR 3 | XP_016860883.1 | |||
XM_017005395.1 | 8314 | UTR 3 | XP_016860884.1 |
Set Membership: |
HapMap |