Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATGGAAGAAAGAACAATAACT[A/C]TAGAAATCCCTGAAGTTCTGAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300609 | ||||||||||||||||||||
Literature Links: |
MSL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSL3 - male-specific lethal 3 homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193270.2 | 595 | Missense Mutation | ATA,CTA | I,L 162 | NP_001180199.1 | |
NM_001282174.1 | 595 | Missense Mutation | ATA,CTA | I,L 25 | NP_001269103.1 | |
NM_006800.3 | 595 | Missense Mutation | ATA,CTA | I,L 8 | NP_006791.2 | |
NM_078628.1 | 595 | Intron | NP_523352.1 | |||
NM_078629.3 | 595 | Missense Mutation | ATA,CTA | I,L 174 | NP_523353.2 |